Assay Details
| Gene Symbol: Zfp449 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030139.4 | NP_084415.1 | 2 - 3 | 482 | 105 | |
| GenBank mRNA | AK020600.1 | - | 2 - 3 | 475 | 105 | |
| AK036596.1 | - | 2 - 3 | 483 | 105 | ||
| AK142888.1 | - | 2 - 3 | 486 | 105 | ||
| BC082594.1 | - | 1 - 2 | 372 | 30642886 | 105 | |
| BC145936.1 | - | 1 - 2 | 372 | 40131239 | 105 | |
| BC145938.1 | - | 1 - 2 | 372 | 40131241 | 105 | |
More Information
Updated Mapping Information
Set Membership: |
|

Zfp449
Genomic Map