Assay Details
| Gene Symbol: Brpf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001282126.1 | NP_001269055.1 | 11 - 12 | 3846 | 79 | |
| NM_001282127.1 | NP_001269056.1 | 12 - 13 | 3509 | 79 | ||
| NM_001282128.1 | NP_001269057.1 | 12 - 13 | 3759 | 79 | ||
| NM_030178.2 | NP_084454.1 | 11 - 12 | 3611 | 79 | ||
| GenBank mRNA | AK019522.1 | - | 6 - 7 | 1136 | 79 | |
| AK156375.1 | - | 11 - 12 | 3794 | 79 | ||
| BC005647.1 | - | 10 - 11 | 1781 | 79 | ||
| BC031594.1 | - | 9 - 10 | 2535 | 79 | ||
| BC046521.1 | - | 11 - 12 | 3551 | 5363697 | 79 | |
| BC138361.1 | - | 12 - 13 | 3333 | 8861381 | 79 | |
| BC145256.1 | - | 12 - 13 | 3583 | 9053809 | 79 | |
| DQ288860.1 | - | 7 - 8 | 2199 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Brpf1
Genomic Map