Assay Details
| Gene Symbol: Brpf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001282127.1 | NP_001269056.1 | 7 - 8 | 2501 | 76 | |
| NM_001282128.1 | NP_001269057.1 | 7 - 8 | 2751 | 76 | ||
| NM_030178.2 | NP_084454.1 | 7 - 8 | 2501 | 76 | ||
| GenBank mRNA | AK019522.1 | - | 1 - 2 | 128 | 76 | |
| AK043372.1 | - | 7 - 8 | 2734 | 76 | ||
| BC005647.1 | - | 5 - 6 | 773 | 76 | ||
| BC031594.1 | - | 5 - 6 | 1425 | 76 | ||
| BC046521.1 | - | 7 - 8 | 2441 | 5363697 | 76 | |
| BC138361.1 | - | 7 - 8 | 2325 | 8861381 | 76 | |
| BC145256.1 | - | 7 - 8 | 2575 | 9053809 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Brpf1
Genomic Map