Assay Details
| Gene Symbol: Brpf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001282126.1 | NP_001269055.1 | 8 - 9 | 3055 | 77 | |
| NM_001282127.1 | NP_001269056.1 | 8 - 9 | 2820 | 77 | ||
| NM_001282128.1 | NP_001269057.1 | 8 - 9 | 3070 | 77 | ||
| NM_030178.2 | NP_084454.1 | 8 - 9 | 2820 | 77 | ||
| GenBank mRNA | AK019522.1 | - | 2 - 3 | 447 | 77 | |
| AK156375.1 | - | 8 - 9 | 3003 | 77 | ||
| BC005647.1 | - | 6 - 7 | 1092 | 77 | ||
| BC031594.1 | - | 6 - 7 | 1744 | 77 | ||
| BC046521.1 | - | 8 - 9 | 2760 | 5363697 | 77 | |
| BC138361.1 | - | 8 - 9 | 2644 | 8861381 | 77 | |
| DQ288860.1 | - | 3 - 4 | 1510 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Brpf1
Genomic Map