Assay Details
| Gene Symbol: Dgkz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166597.1 | NP_001160069.1 | 15 - 16 | 2046 | 71 | |
| NM_138306.2 | NP_612179.2 | 14 - 15 | 1445 | 71 | ||
| GenBank mRNA | AK087503.1 | - | 15 - 16 | 2047 | 71 | |
| AK170936.1 | - | 11 - 12 | 1068 | 71 | ||
| BC049228.1 | - | 14 - 15 | 1481 | 3668704 | 71 | |
| BQ443200.1 | - | 5 - 6 | 449 | 71 | ||
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | AK088841.1 | - | Chr. - - [-] on Build | 71 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Dgkz
Genomic Map