Assay Details
| Gene Symbol: Dgkz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166597.1 | NP_001160069.1 | 10 - 11 | 1550 | 138 | |
| NM_138306.2 | NP_612179.2 | 9 - 10 | 949 | 138 | ||
| GenBank mRNA | AK087503.1 | - | 10 - 11 | 1551 | 138 | |
| AK170936.1 | - | 6 - 7 | 572 | 138 | ||
| BC049228.1 | - | 9 - 10 | 985 | 3668704 | 138 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dgkz
Genomic Map