Assay Details
| Gene Symbol: Brip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178309.2 | NP_840094.1 | 7 - 8 | 1039 | 96 | |
| GenBank mRNA | AK033205.1 | - | 7 - 8 | 1011 | 96 | |
| AK051878.1 | - | 7 - 8 | 1040 | 96 | ||
| AK145709.1 | - | 7 - 8 | 1040 | 96 | ||
| AK169440.1 | - | 7 - 8 | 986 | 96 | ||
| BC094252.1 | - | 7 - 8 | 988 | 6401109 | 96 | |
More Information
Updated Mapping Information
Set Membership: |
|

Brip1
Genomic Map