Assay Details
| Gene Symbol: Dmwd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010058.2 | NP_034188.2 | 2 - 3 | 684 | 86 | |
| GenBank mRNA | AK086899.1 | - | 1 - 2 | 244 | 86 | |
| BC089027.1 | - | 2 - 3 | 916 | 30615314 | 86 | |
| CF169673.1 | - | 2 - 3 | 347 | 86 | ||
| S60312.1 | - | 2 - 3 | 324 | 86 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24991258 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24991259 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24991256 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24991257 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2013414 | BC089027.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2013414 | S60312.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2013414 | NM_010058.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2013414 | AK086899.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2013414 | BC031193.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 20585 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 651430 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 356164 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 227005 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 133912 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P147195 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2013414 | BC089027.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2013414 | S60312.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2013414 | NM_010058.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2013414 | AK086899.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2013414 | BC031193.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24991262 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24991267 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24991281 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24991276 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24991306 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24991302 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24991289 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24991287 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24991273 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24991291 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24991269 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24991259 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24991299 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24991254 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24991251 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24991294 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24991310 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24991290 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24991298 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24991304 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24991271 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24991275 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24991279 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24991283 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24991305 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24991261 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24991308 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24991258 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24991278 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24991266 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24991282 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24991255 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24991285 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24991264 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24991296 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24991252 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24991270 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24991265 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06134084 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P147195 | - | - |
| Whole Mouse Genome G4122A | A_51_P483946 | BC089027.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P483946 | S60312.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P483946 | NM_010058.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P483946 | AK086899.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P483946 | BC031193.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P130538 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P147195 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06141205 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06141208 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06527093 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06527091 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06141209 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06141207 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06095806 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06141204 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24991259 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24991258 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24991257 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24991256 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24991379 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24991358 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24991320 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24991350 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24991331 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24991326 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24991270 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24991382 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24991289 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24991259 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24991375 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24991302 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24991291 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24991273 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24991380 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24991321 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24991325 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24991308 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24991329 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24991251 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24991258 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24991317 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24991346 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24991276 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24991265 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24991279 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24991356 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24991261 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24991296 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24991282 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24991314 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24991323 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24991333 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24991290 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24991298 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24991341 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24991268 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24991340 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24991312 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24991285 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24991305 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24991269 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24991299 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24991369 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24991368 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24991364 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24991306 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24991294 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24991332 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24991254 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24991287 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24991352 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24991281 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24991342 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24991310 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24991360 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24991284 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24991354 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24991262 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24991370 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24991304 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24991278 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24991283 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24991348 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24991343 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24991345 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06274028 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06274034 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06141206 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24991269 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06502501 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24991279 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06274042 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24991278 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24991296 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24991306 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24991299 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06193370 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06274040 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06529510 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24991264 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24991298 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24991305 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24991254 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24991281 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24991256 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24991268 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06141205 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24991275 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24991291 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06141204 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24991252 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06193326 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24991284 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06274025 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06141209 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06502506 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24991310 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24991301 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24991285 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06274039 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24991276 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10550571 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10550569 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10550566 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10550568 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10550567 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10550565 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10550572 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10550570 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10550573 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1447642_PM_x_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1435505_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1447641_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10550564 | - | - |
| Mouse Genome 430 2.0 Array | 1447641_at | - | - |
| Mouse Genome 430 2.0 Array | 1447641_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1447642_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1435505_at | - | - |
| Mouse Genome 430 2.0 Array | 1435505_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1447642_x_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 13401_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 13401_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700002109.mm.1 | TC0700000284.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700002109.mm.1 | NM_010058.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700002109.mm.1 | NM_010058.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700002104.mm.1 | TC0700000284.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700002106.mm.1 | TC0700000284.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700002105.mm.1 | TC0700000284.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700002102.mm.1 | TC0700000284.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700002102.mm.1 | NM_010058.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700002102.mm.1 | NM_010058.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700002110.mm.1 | TC0700000284.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700002110.mm.1 | NM_010058.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700002110.mm.1 | NM_010058.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700002111.mm.1 | TC0700000284.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700002111.mm.1 | NM_010058.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700002111.mm.1 | NM_010058.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700002100.mm.1 | TC0700000284.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700002100.mm.1 | NM_010058.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700002100.mm.1 | NM_010058.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700002108.mm.1 | TC0700000284.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700002108.mm.1 | NM_010058.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700002108.mm.1 | NM_010058.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700002103.mm.1 | TC0700000284.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700002107.mm.1 | TC0700000284.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700002107.mm.1 | NM_010058.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700002107.mm.1 | NM_010058.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700002112.mm.1 | TC0700000284.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700002112.mm.1 | NM_010058.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700002112.mm.1 | NM_010058.2 | Refseq |
| Mouse Expression Set 430 | 1435505_PM_at | - | - |
| Mouse Expression Set 430 | 1447641_PM_at | - | - |
| Mouse Expression Set 430 | 1435505_at | - | - |
| Mouse Expression Set 430 | 1447642_x_at | - | - |
| Mouse Expression Set 430 | 1447641_at | - | - |
| Mouse Expression Set 430 | 1447642_PM_x_at | - | - |
| Murine Genome U74v2 Set | 96193_at | - | - |
| Murine Genome U74v2 Set | 96193_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1435505_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1447642_x_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1447641_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1435505_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1447641_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1447642_PM_x_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 514245 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 1690047 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1690047 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 770019 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000770019 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001690047 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2711355 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1253447 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1253447 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2711355 | - | - |
| mouse-6 v1.1 (Illumina) | scl33012.5.1_34-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0004.1_5-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1690047 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 770019 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0004.1_5-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl33012.5.1_34-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1690047 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 770019 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2711355 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1253447 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2914010 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1253447 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2711355 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2914010 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl33012.5.1_34-S | - | - |

Dmwd
Genomic Map