Assay Details
| Gene Symbol: Nabp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028696.3 | NP_082972.2 | 2 - 3 | 714 | 88 | |
| GenBank mRNA | AK015312.1 | - | 2 - 3 | 234 | 88 | |
| AK028886.1 | - | 2 - 3 | 715 | 88 | ||
| AK028936.1 | - | 1 - 2 | 590 | 88 | ||
| AK031015.1 | - | 1 - 2 | 924 | 88 | ||
| AK049396.1 | - | 2 - 3 | 715 | 88 | ||
| AK051607.1 | - | 2 - 3 | 715 | 88 | ||
| AK077226.1 | - | 2 - 3 | 168 | 88 | ||
| AK079434.1 | - | 2 - 3 | 819 | 88 | ||
| AK083915.1 | - | 2 - 3 | 715 | 88 | ||
| AK163613.1 | - | 1 - 2 | 923 | 88 | ||
| AK166300.1 | - | 2 - 3 | 715 | 88 | ||
| AK167541.1 | - | 2 - 3 | 167 | 88 | ||
| AK167652.1 | - | 2 - 3 | 716 | 88 | ||
| AY880264.1 | - | 2 - 3 | 715 | 88 | ||
| BC027371.1 | - | 1 - 2 | 144 | 88 | ||
| BC095967.1 | - | 2 - 3 | 380 | 6486629 | 88 | |
| BC131923.1 | - | 2 - 3 | 260 | 40130200 | 88 | |
| BC132532.1 | - | 2 - 3 | 260 | 40130809 | 88 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK028936.1, AK163613.1 transcripts.
Set Membership: |
|

Nabp1
Genomic Map