Assay Details
| Gene Symbol: Fam20c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030565.6 | NP_085042.2 | 1 - 2 | 1149 | 62 | |
| GenBank mRNA | AK154810.1 | - | 1 - 2 | 1145 | 62 | |
| AK220286.1 | - | 1 - 2 | 666 | 62 | ||
| AY778962.1 | - | 1 - 2 | 1130 | 62 | ||
| BC025814.1 | - | 1 - 2 | 1100 | 62 | ||
| BC025826.1 | - | 1 - 2 | 1130 | 62 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC025814.1, BC025826.1 transcripts.
Set Membership: |
|

Fam20c
Genomic Map