Assay Details
| Gene Symbol: Pabpc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130881.2 | NP_570951.2 | 10 - 11 | 2155 | 158 | |
| GenBank mRNA | AK135962.1 | - | 1620 | 159 | ||
| AK160589.1 | - | 3 - 4 | 237 | 158 | ||
| AK162172.1 | - | 2 - 3 | 237 | 158 | ||
| BC003283.1 | - | 10 - 11 | 1702 | 3582181 | 158 | |
| BC056432.1 | - | 10 - 11 | 2155 | 5696369 | 158 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Pabpc4
Genomic Map