Assay Details
Gene Symbol: Adamtsl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001301705.1 | NP_001288634.1 | 11 - 12 | 2229 | 92 | |
NM_144899.3 | NP_659148.2 | 11 - 12 | 2111 | 92 | ||
GenBank mRNA | AK155481.1 | - | 11 - 12 | 2162 | 92 | |
AY158701.1 | - | 11 - 12 | 2052 | 92 | ||
BC117925.1 | - | 10 - 11 | 2024 | 40097608 | 92 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
