Assay Details
Gene Symbol: Ifi27l2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145449.2 | NP_663424.1 | 4 - 5 | 447 | 165 | |
GenBank mRNA | AK007590.1 | - | 2 - 3 | 338 | 165 | |
BC021795.1 | - | 4 - 5 | 447 | 4217067 | 165 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2085776 | NM_145449.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2085776 | BC021795.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2085779 | NM_145449.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2085779 | AK007590.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2085779 | BC021795.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 339466 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 864658 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 235940 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 735551 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 10770 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P153877 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2085779 | NM_145449.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2085779 | AK007590.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2085779 | BC021795.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2085776 | NM_145449.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2085776 | BC021795.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28697697 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28697694 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28697691 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28697711 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P153877 | - | - |
Whole Mouse Genome G4122A | A_51_P391432 | NM_145449.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P391432 | AK007590.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P391432 | BC021795.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P153877 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10340654 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10340658 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09780065 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28697660 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28697636 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28697647 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28697621 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28697709 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28697638 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28697635 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28697652 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28697632 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28697654 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28697625 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28697687 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28697619 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28697659 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28697622 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28697688 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28697627 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28697626 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28697649 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28697690 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28697663 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28697623 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28697694 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28697661 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28697665 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28697646 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28697630 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28697624 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28697683 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28697675 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28697688 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10340658 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10171617 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28697687 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28697674 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28697692 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10340651 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28697690 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28697711 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10402359 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10402354 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10402358 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10402355 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10402356 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10402357 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1425120_PM_x_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10402353 | - | - |
Mouse Genome 430 2.0 Array | 1425120_x_at | - | - |
Mouse Genome 430 2.0 Array | 1425120_PM_x_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 217845_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 217845_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1200015766.mm.1 | NM_145449.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1200015766.mm.1 | NM_145449.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1200015766.mm.1 | TC1200002270.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1200015765.mm.1 | NM_145449.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1200015765.mm.1 | NM_145449.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1200015765.mm.1 | TC1200002270.mm.1 | MTA TC |
Mouse Expression Set 430 | 1425120_PM_x_at | - | - |
Mouse Expression Set 430 | 1425120_x_at | - | - |
Murine Genome U74v2 Set | 165944_at | - | - |
Murine Genome U74v2 Set | 168934_at | - | - |
Murine Genome U74v2 Set | 165944_PM_at | - | - |
Murine Genome U74v2 Set | 168934_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1425120_PM_x_at | - | - |
Mouse Genome 430A 2.0 Array | 1425120_x_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1425120_PM_x_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1425120_x_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 644676 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 5570048 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0005570048 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1232995 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1232995 | - | - |
mouse-6 v1.1 (Illumina) | scl42110.7.200_45-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 5570048 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl42110.7.200_45-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5570048 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1232995 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1232995 | - | - |
