Assay Details
| Gene Symbol: Spag11a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153115.1 | NP_694755.1 | 1 - 2 | 111 | 153 | |
| GenBank mRNA | AB089183.1 | - | 1 - 2 | 111 | 153 | |
| AF384173.1 | - | 1 - 2 | 74 | 153 | ||
| AK020333.1 | - | 1 - 2 | 115 | 153 | ||
| BC120808.1 | - | 1 - 2 | 76 | 40129731 | 153 | |
| BC120810.1 | - | 1 - 2 | 76 | 40129733 | 153 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P438485 | NM_153115.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P438485 | AB089183.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P438485 | AF384173.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P438485 | AK020333.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P438485 | AY552529.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P438485 | NM_153115.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P438485 | AB089183.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P438485 | AF384173.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P438485 | AK020333.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P438485 | AY552529.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 364861 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 299936 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 305278 | AK009711.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 305278 | AK135812.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 800266 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 145205 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 500949 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 132082 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 81542 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 13940 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P133525 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P438485 | NM_153115.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P438485 | AB089183.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P438485 | AF384173.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P438485 | AK020333.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P438485 | AY552529.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P438485 | NM_153115.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P438485 | AB089183.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P438485 | AF384173.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P438485 | AK020333.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P438485 | AY552529.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25702262 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25702232 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25702250 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25702231 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25702247 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25702240 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25702261 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25702236 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25702238 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25702234 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25702255 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25702253 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25702244 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25702233 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25702263 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25702235 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25702252 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25702242 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25702256 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25702230 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25702260 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25702248 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25702259 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25702258 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25702254 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06443222 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P133525 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06443208 | - | - |
| Whole Mouse Genome G4122A | A_52_P438485 | NM_153115.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P438485 | AB089183.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P438485 | AF384173.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P438485 | AK020333.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P438485 | AY552529.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P438485 | NM_153115.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P438485 | AB089183.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P438485 | AF384173.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P438485 | AK020333.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P438485 | AY552529.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P135128 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P133525 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P06443222 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07378413 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07378410 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07378412 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07378405 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07378406 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07378416 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702233 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702231 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702219 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702335 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702272 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702270 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702210 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702294 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702338 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702230 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702250 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702253 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702216 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702274 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702247 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702286 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702269 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702290 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702218 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702221 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702255 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702320 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702284 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702316 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702282 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702227 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702330 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702310 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702291 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702235 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702267 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702244 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702329 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702248 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702297 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702266 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702317 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702258 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702324 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702254 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702296 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702304 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702220 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702256 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702252 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702223 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702242 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702312 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702238 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702234 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702318 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702303 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702327 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702277 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702260 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702309 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702279 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702314 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702240 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702232 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702213 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702262 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702298 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702333 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25702264 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25702269 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25702294 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25702247 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25702296 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25702242 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25702230 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25702254 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07425798 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06669169 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25702231 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25702260 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07425804 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07378410 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25702240 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06767048 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25702233 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25702274 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07374717 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25702252 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07208692 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07297282 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25702250 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07297283 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25702267 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25702282 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25702270 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06669224 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07052282 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25702232 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25702277 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07052281 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25702238 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25702258 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25702297 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25702290 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25702266 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25702302 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25702262 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07026531 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25702210 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25702286 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25702255 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25702253 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25702303 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25702298 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25702284 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25702244 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25702234 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25702291 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10570603 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10570602 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10570600 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10570591 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10570601 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10570590 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10570599 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0800001574.mm.1 | NM_153115.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800001574.mm.1 | NM_153115.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800001574.mm.1 | TC0800000178.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800001576.mm.1 | NM_153115.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800001576.mm.1 | NM_153115.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800001576.mm.1 | TC0800000178.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800001575.mm.1 | NM_153115.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800001575.mm.1 | NM_153115.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800001575.mm.1 | TC0800000178.mm.1 | MTA TC |
| Murine Genome U74v2 Set | 168506_PM_i_at | - | - |
| Murine Genome U74v2 Set | 168506_i_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 308239 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 100540309 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2480017 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 100540309 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002480017 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1248179 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1260418 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1248179 | - | - |
| mouse-6 v1.1 (Illumina) | GI_23346552-S | - | - |
| mouse-6 v1.1 (Illumina) | scl078128.1_11-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2480017 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 100540309 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl078128.1_11-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 100540309 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2480017 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1248179 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1248179 | - | - |

Spag11a
Genomic Map