Assay Details
| Gene Symbol: Ppp1r10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163818.1 | NP_001157290.1 | 2 - 3 | 635 | 156 | |
| GenBank mRNA | AJ504718.1 | - | 2 - 3 | 641 | 156 | |
| AK053183.1 | - | 3 - 4 | 654 | 156 | ||
| BC030903.1 | - | 2 - 3 | 642 | 156 | ||
| BC052059.1 | - | 2 - 3 | 638 | 5702171 | 156 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ppp1r10
Genomic Map