Assay Details
| Gene Symbol: Sirt5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178848.3 | NP_849179.1 | 2 - 3 | 295 | 73 | |
| GenBank mRNA | AK002609.1 | - | 3 - 4 | 325 | 73 | |
| BC031770.1 | - | 2 - 3 | 295 | 4014119 | 73 | |
| BC087898.1 | - | 3 - 4 | 317 | 30254686 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sirt5
Genomic Map