Assay Details
| Gene Symbol: Chchd7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181391.4 | NP_852056.1 | 1 - 2 | 246 | 96 | |
| NR_104354.1 | 1 - 2 | 246 | 96 | |||
| NR_104355.1 | 1 - 2 | 246 | 96 | |||
| GenBank mRNA | AK007844.1 | - | 1 - 2 | 221 | 96 | |
| AK013200.1 | - | 1 - 2 | 219 | 96 | ||
| AK145105.1 | - | 1 - 2 | 96 | |||
| AK169018.1 | - | 1 - 2 | 221 | 96 | ||
| BC030366.1 | - | 1 - 2 | 5370748 | 95 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK007844.1, AK013200.1, AK145105.1, AK169018.1, BC030366.1, NM_181391.4 transcripts.
Set Membership: |
|

Chchd7
Genomic Map