Assay Details
| Gene Symbol: Cntnap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016782.2 | NP_058062.2 | 13 - 14 | 2206 | 58 | |
| GenBank mRNA | AK154468.1 | - | 2967 | 58 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cntnap1
Genomic Map