Assay Details
| Gene Symbol: Myt1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001093775.1 | NP_001087244.1 | 14 - 15 | 2943 | 191 | |
| NM_001093776.1 | NP_001087245.1 | 14 - 15 | 2849 | 191 | ||
| NM_001093778.1 | NP_001087247.1 | 14 - 15 | 2934 | 191 | ||
| NM_008666.3 | NP_032692.2 | 14 - 15 | 2937 | 191 | ||
| GenBank mRNA | AB093283.1 | - | 11 - 12 | 2296 | 191 | |
| AB212898.1 | - | 13 - 14 | 2541 | 191 | ||
| AF004295.1 | - | 14 - 15 | 2753 | 191 | ||
| AK049967.1 | - | 5 - 6 | 778 | 191 | ||
| AK076608.1 | - | 2 - 3 | 374 | 191 | ||
| BC079651.1 | - | 14 - 15 | 2818 | 191 | ||
| BC094438.1 | - | 10 - 11 | 2174 | 30692780 | 191 | |
| BC131677.1 | - | 13 - 14 | 2538 | 40111036 | 191 | |
| BQ876229.1 | - | 1 - 2 | 71 | 191 | ||
| U86338.1 | - | 14 - 15 | 2665 | 191 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Myt1l
Genomic Map