| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1100024900.mm.1 |
NM_009012.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100024900.mm.1 |
NM_009012.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100024900.mm.1 |
TC1100002740.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100024901.mm.1 |
NM_009012.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100024901.mm.1 |
NM_009012.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100024901.mm.1 |
TC1100002740.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100024902.mm.1 |
NM_009012.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100024902.mm.1 |
NM_009012.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100024902.mm.1 |
TC1100002740.mm.1 |
MTA TC |