Assay Details
| Gene Symbol: Prkcz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039079.2 | NP_001034168.1 | 4 - 5 | 634 | 102 | |
| NM_008860.3 | NP_032886.2 | 7 - 8 | 834 | 102 | ||
| GenBank mRNA | AB110830.1 | - | 4 - 5 | 594 | 102 | |
| AK042562.1 | - | 4 - 5 | 592 | 102 | ||
| AK049959.1 | - | 4 - 5 | 618 | 102 | ||
| AK147300.1 | - | 4 - 5 | 452 | 102 | ||
| BC072590.1 | - | 4 - 5 | 634 | 102 | ||
| BC139761.1 | - | 7 - 8 | 651 | 40109623 | 102 | |
| M94632.1 | - | 7 - 8 | 651 | 102 | ||
More Information
Set Membership: |
|

Prkcz
Genomic Map