| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0800020305.mm.1 |
NM_033327.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800020305.mm.1 |
NM_033327.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800020305.mm.1 |
TC0800002648.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800020301.mm.1 |
NM_033327.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800020301.mm.1 |
NM_033327.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800020301.mm.1 |
TC0800002648.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800020302.mm.1 |
NM_033327.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800020302.mm.1 |
NM_033327.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800020302.mm.1 |
TC0800002648.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800020303.mm.1 |
NM_033327.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800020303.mm.1 |
NM_033327.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800020303.mm.1 |
TC0800002648.mm.1 |
MTA TC |