Assay Details
| Gene Symbol: Cdk4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009870.3 | NP_034000.1 | 2 - 3 | 387 | 62 | |
| GenBank mRNA | AK003804.1 | - | 2 - 3 | 354 | 62 | |
| AK006918.1 | - | 2 - 3 | 354 | 62 | ||
| AK011871.1 | - | 2 - 3 | 375 | 62 | ||
| AK012064.1 | - | 2 - 3 | 356 | 62 | ||
| AK019380.1 | - | 2 - 3 | 341 | 62 | ||
| AK078440.1 | - | 2 - 3 | 341 | 62 | ||
| AK146107.1 | - | 2 - 3 | 354 | 62 | ||
| AK153465.1 | - | 2 - 3 | 387 | 62 | ||
| BC031599.1 | - | 2 - 3 | 376 | 62 | ||
| BC046336.1 | - | 2 - 3 | 387 | 6312184 | 62 | |
| BC052694.1 | - | 2 - 3 | 382 | 5714971 | 62 | |
| L01640.1 | - | 1 - 2 | 214 | 62 | ||
More Information
Set Membership: |
|

Cdk4
Genomic Map