Assay Details
| Gene Symbol: Cdc42bpa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033285.1 | NP_001028457.1 | 3 - 4 | 378 | 66 | |
| GenBank mRNA | AK136237.1 | - | 5 - 6 | 575 | 66 | |
| AK138649.1 | - | 4 - 5 | 867 | 66 | ||
| AK162174.1 | - | 3 - 4 | 276 | 66 | ||
| BC158017.1 | - | 3 - 4 | 378 | 9007323 | 66 | |
| BC158095.1 | - | 3 - 4 | 378 | 9007382 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cdc42bpa
Genomic Map