Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100017322.mm.1 |
NM_011943.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100017322.mm.1 |
NM_011943.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100017322.mm.1 |
TC1100001841.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100017323.mm.1 |
NM_011943.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100017323.mm.1 |
NM_011943.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100017323.mm.1 |
TC1100001841.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100017324.mm.1 |
NM_011943.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100017324.mm.1 |
NM_011943.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100017324.mm.1 |
TC1100001841.mm.1 |
MTA TC |