Assay Details
| Gene Symbol: Hdgf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008231.4 | NP_032257.3 | 5 - 6 | 1008 | 76 | |
| GenBank mRNA | AF251787.1 | - | 5 - 6 | 921 | 76 | |
| AK005359.1 | - | 5 - 6 | 786 | 76 | ||
| AK017863.1 | - | 5 - 6 | 962 | 76 | ||
| AK029475.1 | - | 5 - 6 | 752 | 76 | ||
| AK076021.1 | - | 5 - 6 | 796 | 76 | ||
| BC005713.1 | - | 5 - 6 | 918 | 2609261 | 76 | |
| BC021654.1 | - | 5 - 6 | 936 | 4163897 | 76 | |
| D63707.1 | - | 5 - 6 | 762 | 76 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Hdgf
Genomic Map