Assay Details
| Gene Symbol: Rragb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004154.2 | NP_001004154.1 | 4 - 5 | 830 | 113 | |
| GenBank mRNA | BC069180.1 | - | 4 - 5 | 788 | 6810543 | 113 |
| BC070407.1 | - | 4 - 5 | 807 | 6808979 | 113 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rragb
Genomic Map