Assay Details
| Gene Symbol: Pde4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001177980.1 | NP_001171451.1 | 3 - 4 | 1086 | 63 | |
| NM_001177981.1 | NP_001171452.1 | 3 - 4 | 626 | 63 | ||
| NM_001177982.1 | NP_001171453.1 | 3 - 4 | 362 | 63 | ||
| NM_019840.2 | NP_062814.2 | 8 - 9 | 1251 | 63 | ||
| GenBank mRNA | AF326555.1 | - | 8 - 9 | 991 | 63 | |
| AF326556.1 | - | 8 - 9 | 991 | 63 | ||
| AK051102.1 | - | 3 - 4 | 627 | 63 | ||
| AK150868.1 | - | 3 - 4 | 592 | 63 | ||
| AK161341.1 | - | 3 - 4 | 363 | 63 | ||
| BC007155.1 | - | 3 - 4 | 433 | 63 | ||
| BC023751.1 | - | 3 - 4 | 1062 | 63 | ||
| BC071259.1 | - | 3 - 4 | 602 | 5368075 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pde4b
Genomic Map