| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0700019011.mm.1 |
TC0700002105.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700019011.mm.1 |
NM_001081370.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700019011.mm.1 |
NM_001081370.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700019011.mm.1 |
NM_001113373.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700019011.mm.1 |
NM_001113373.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700019013.mm.1 |
TC0700002105.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700019013.mm.1 |
NM_001081370.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700019013.mm.1 |
NM_001081370.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700019013.mm.1 |
NM_001113373.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700019013.mm.1 |
NM_001113373.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700019004.mm.1 |
TC0700002105.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700019004.mm.1 |
NM_001081370.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700019004.mm.1 |
NM_001081370.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700019004.mm.1 |
NM_001113373.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700019004.mm.1 |
NM_001113373.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700019005.mm.1 |
TC0700002105.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700019005.mm.1 |
NM_001081370.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700019005.mm.1 |
NM_001081370.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700019005.mm.1 |
NM_001113373.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700019005.mm.1 |
NM_001113373.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700019010.mm.1 |
TC0700002105.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700019010.mm.1 |
NM_001081370.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700019010.mm.1 |
NM_001081370.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700019010.mm.1 |
NM_001113373.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700019010.mm.1 |
NM_001113373.2 |
Refseq |