Assay Details
| Gene Symbol: Pigl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039536.2 | NP_001034625.1 | 4 - 5 | 492 | 73 | |
| GenBank mRNA | AK158149.1 | - | 4 - 5 | 493 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pigl
Genomic Map