Assay Details
| Gene Symbol: Ppp3cb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008914.2 | NP_032940.1 | 3 - 4 | 536 | 131 | |
| GenBank mRNA | AB221678.1 | - | 3 - 4 | 403 | 131 | |
| AK135203.1 | - | 3 - 4 | 350 | 131 | ||
| BC037668.1 | - | 3 - 4 | 359 | 131 | ||
| BC066000.1 | - | 3 - 4 | 507 | 5699179 | 131 | |
| M81483.1 | - | 3 - 4 | 373 | 131 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ppp3cb
Genomic Map