Assay Details
| Gene Symbol: Tle4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF229633.1 | - | 4 - 5 | 264 | 103 | |
| AK134989.1 | - | 4 - 5 | 895 | 103 | ||
| BC086631.1 | - | 5 - 6 | 808 | 103 | ||
| BQ769921.1 | - | 4 - 5 | 315 | 103 | ||
More Information
Set Membership: |
|

Tle4
Genomic Map