Assay Details
| Gene Symbol: Slc9a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177084.3 | NP_796058.1 | 3 - 4 | 1274 | 91 | |
| GenBank mRNA | AF139195.1 | - | 1 - 2 | 76 | 91 | |
| AK085681.1 | - | 3 - 4 | 1300 | 91 | ||
| AK141534.1 | - | 3 - 4 | 1275 | 91 | ||
| AK170522.1 | - | 1 - 2 | 113 | 91 | ||
| BC120543.1 | - | 3 - 4 | 1192 | 40129466 | 91 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc9a4
Genomic Map