Assay Details
| Gene Symbol: Grik3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081097.2 | NP_001074566.1 | 10 - 11 | 1532 | 73 | |
| GenBank mRNA | AF245444.1 | - | 2 - 3 | 327 | 73 | |
| BC118004.1 | - | 10 - 11 | 1527 | 73 | ||
| CF744155.1 | - | 1 - 2 | 185 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Grik3
Genomic Map