Assay Details
| Gene Symbol: Epha3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010140.3 | NP_034270.1 | 12 - 13 | 2245 | 81 | |
| GenBank mRNA | AK045219.1 | - | 12 - 13 | 2246 | 81 | |
| AK084921.1 | - | 12 - 13 | 2225 | 81 | ||
| AK132335.1 | - | 12 - 13 | 2194 | 81 | ||
| BC060164.1 | - | 12 - 13 | 2222 | 82 | ||
| BC093483.1 | - | 2246 | 81 | |||
| BC138641.1 | - | 12 - 13 | 2168 | 8861661 | 81 | |
| BC138643.1 | - | 12 - 13 | 2168 | 8861663 | 81 | |
| BC145349.1 | - | 12 - 13 | 2165 | 9053905 | 81 | |
| M68513.1 | - | 12 - 13 | 2220 | 81 | ||
More Information
Set Membership: |
|

Epha3
Genomic Map