Assay Details
| Gene Symbol: Epha3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010140.3 | NP_034270.1 | 9 - 10 | 1875 | 86 | |
| GenBank mRNA | AK045219.1 | - | 9 - 10 | 1876 | 86 | |
| AK084921.1 | - | 9 - 10 | 1855 | 86 | ||
| AK132335.1 | - | 9 - 10 | 1824 | 86 | ||
| BC053479.1 | - | 5 - 6 | 767 | 86 | ||
| BC060164.1 | - | 9 - 10 | 1852 | 86 | ||
| BC093483.1 | - | 1876 | 86 | |||
| BC138641.1 | - | 9 - 10 | 1798 | 8861661 | 86 | |
| BC138643.1 | - | 9 - 10 | 1798 | 8861663 | 86 | |
| BC145349.1 | - | 9 - 10 | 1795 | 9053905 | 86 | |
| M68513.1 | - | 9 - 10 | 1850 | 86 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Epha3
Genomic Map