Assay Details
| Gene Symbol: Rnasel | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011882.2 | NP_036012.1 | 7 - 8 | 2511 | 126 | |
| GenBank mRNA | AF281045.1 | - | 5 - 6 | 2055 | 126 | |
| AK053125.1 | - | 6 - 7 | 2210 | 126 | ||
| AK140815.1 | - | 7 - 8 | 2511 | 126 | ||
| AK172300.1 | - | 4 - 5 | 514 | 126 | ||
| BC138013.1 | - | 4 - 5 | 1928 | 8861032 | 126 | |
| JX443419.1 | - | 4 - 5 | 126 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Rnasel
Genomic Map