Assay Details
| Gene Symbol: Cfap36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025740.3 | NP_080016.1 | 2 - 3 | 320 | 153 | |
| GenBank mRNA | AK010468.1 | - | 2 - 3 | 347 | 153 | |
| AK016479.1 | - | 2 - 3 | 356 | 153 | ||
| AK075858.1 | - | 2 - 3 | 321 | 153 | ||
| AK219590.1 | - | 2 - 3 | 110 | 153 | ||
| BC005731.1 | - | 2 - 3 | 290 | 3595965 | 153 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cfap36
Genomic Map