Assay Details
| Gene Symbol: Rps6ka6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025949.3 | NP_080225.2 | 17 - 18 | 1747 | 153 | |
| GenBank mRNA | AK012150.2 | - | 6 - 7 | 499 | 151 | |
| AK049349.1 | - | 15 - 16 | 1498 | 153 | ||
| BC054113.1 | - | 15 - 16 | 1420 | 30015425 | 153 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rps6ka6
Genomic Map