Assay Details
| Gene Symbol: Mcm10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027290.3 | NP_081566.2 | 7 - 8 | 1151 | 66 | |
| GenBank mRNA | AK030780.1 | - | 7 - 8 | 1096 | 66 | |
| AK041406.1 | - | 7 - 8 | 1086 | 66 | ||
| AK041749.1 | - | 1 - 2 | 157 | 66 | ||
| BC064314.1 | - | 7 - 8 | 1514 | 66 | ||
| BC094576.1 | - | 7 - 8 | 1151 | 6853824 | 66 | |
| BC120687.1 | - | 6 - 7 | 1060 | 40129610 | 66 | |
| BC120689.1 | - | 6 - 7 | 1060 | 40129612 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mcm10
Genomic Map