Assay Details
| Gene Symbol: Zmiz2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005867.1 | NP_001005867.1 | 15 - 16 | 2131 | 106 | |
| NM_028601.3 | NP_082877.2 | 15 - 16 | 2227 | 106 | ||
| GenBank mRNA | AK048533.1 | - | 121 | 106 | ||
| BC024081.1 | - | 14 - 15 | 1977 | 106 | ||
| BC040745.1 | - | 6 - 7 | 686 | 106 | ||
| BC060652.1 | - | 15 - 16 | 2069 | 30354393 | 106 | |
| BC063069.1 | - | 7 - 8 | 832 | 106 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK048533.1 transcripts.
Set Membership: |
|

Zmiz2
Genomic Map