Assay Details
| Gene Symbol: Sema3d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028882.4 | NP_083158.3 | 11 - 12 | 1733 | 73 | |
| GenBank mRNA | AK028900.1 | - | 11 - 12 | 1515 | 73 | |
| AK031704.1 | - | 11 - 12 | 1345 | 73 | ||
| AK052671.1 | - | 11 - 12 | 1734 | 73 | ||
| AK153839.1 | - | 11 - 12 | 1394 | 73 | ||
| BC138131.1 | - | 11 - 12 | 1276 | 8861150 | 73 | |
More Information
Set Membership: |
|

Sema3d
Genomic Map