Assay Details
| Gene Symbol: Sema3d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028882.4 | NP_083158.3 | 1 - 2 | 512 | 106 | |
| GenBank mRNA | AK028900.1 | - | 1 - 2 | 294 | 106 | |
| AK031704.1 | - | 1 - 2 | 124 | 106 | ||
| AK051165.1 | - | 1 - 2 | 173 | 106 | ||
| AK052671.1 | - | 1 - 2 | 513 | 106 | ||
| AK087013.1 | - | 1 - 2 | 114 | 106 | ||
| AK134412.1 | - | 1 - 2 | 148 | 106 | ||
| AK153839.1 | - | 1 - 2 | 173 | 106 | ||
| BC138131.1 | - | 1 - 2 | 55 | 8861150 | 106 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK028900.1, AK031704.1, AK051165.1, AK052671.1, AK087013.1, AK134412.1, BC138131.1, NM_028882.4 transcripts.
Set Membership: |
|

Sema3d
Genomic Map