Assay Details
| Gene Symbol: Sema3d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028882.4 | NP_083158.3 | 7 - 8 | 1267 | 141 | |
| GenBank mRNA | AK028900.1 | - | 7 - 8 | 1049 | 141 | |
| AK031704.1 | - | 7 - 8 | 879 | 141 | ||
| AK051165.1 | - | 7 - 8 | 928 | 141 | ||
| AK052671.1 | - | 7 - 8 | 1268 | 141 | ||
| AK153839.1 | - | 7 - 8 | 928 | 141 | ||
| BC138131.1 | - | 7 - 8 | 810 | 8861150 | 141 | |
More Information
Set Membership: |
|

Sema3d
Genomic Map