Assay Details
| Gene Symbol: Sema3d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028882.4 | NP_083158.3 | 8 - 9 | 1414 | 142 | |
| GenBank mRNA | AK028900.1 | - | 8 - 9 | 1196 | 142 | |
| AK031704.1 | - | 8 - 9 | 1026 | 142 | ||
| AK051165.1 | - | 8 - 9 | 1075 | 142 | ||
| AK052671.1 | - | 8 - 9 | 1415 | 142 | ||
| AK153839.1 | - | 8 - 9 | 1075 | 142 | ||
| BC138131.1 | - | 8 - 9 | 957 | 8861150 | 142 | |
More Information
Set Membership: |
|

Sema3d
Genomic Map