Assay Details
| Gene Symbol: Sun5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK006225.1 | - | 11 - 12 | 971 | 138 | |
| AY307077.1 | - | 11 - 12 | 990 | 138 | ||
More Information
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2068121 | AY307077.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2068121 | NM_029599.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2068121 | AK006225.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 186256 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 229997 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 613405 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 371384 | AK038584.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 371384 | AK043388.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 371384 | AK043589.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 371384 | NM_178797.3 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 800319 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 267402 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 902534 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 278742 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 109897 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P126245 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P137845 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2068121 | AY307077.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2068121 | NM_029599.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2068121 | AK006225.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21757428 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21757450 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21757454 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21757431 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21757430 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21757433 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21757438 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21757426 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21757418 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21757423 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21757441 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21757421 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21757442 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21757437 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21757444 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21757434 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21757435 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21757464 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21757451 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21757466 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21757448 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21757453 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21757465 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21757424 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21757420 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04745540 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P126245 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P137845 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00530994 | - | - |
| Whole Mouse Genome G4122A | A_51_P474278 | FJ667498.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P474278 | AY307077.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P474278 | NM_029599.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P474278 | AK006225.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P109536 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P126245 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P137845 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01891998 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01885228 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01891974 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01885223 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01495929 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01885227 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01495944 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01495921 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757371 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757451 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757408 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757366 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757467 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757317 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757335 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757327 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757356 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757421 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757500 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757389 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757353 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757373 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757357 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757505 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757321 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757485 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757416 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757424 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757445 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757344 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757426 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757387 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757302 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757475 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757433 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757482 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757476 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757360 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757354 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757442 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757487 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757492 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757397 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757391 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757507 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757434 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757410 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757463 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757346 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757496 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757308 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757472 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757396 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757423 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757431 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757359 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757361 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757310 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757352 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757338 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757394 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757298 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757349 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757312 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757486 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757375 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757307 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757323 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757331 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757474 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757405 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757489 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757483 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757322 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757502 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757495 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757413 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757428 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757490 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757329 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757319 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757380 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757441 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757320 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757435 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757468 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757469 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757465 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757453 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757341 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757481 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757464 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757369 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757418 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757448 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757430 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757400 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757497 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757510 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757377 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757337 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757325 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21757439 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01891994 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01663537 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21757431 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21757423 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21757426 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21757421 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21757441 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21757434 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21757387 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21757384 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21757448 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21757410 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21757413 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21757402 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01441080 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21757428 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21757392 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01663532 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21757433 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01441061 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21757394 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21757424 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21757405 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21757442 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21757386 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01441065 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21757397 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21757396 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21757389 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21757445 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21757435 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01441074 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21757391 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21757407 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21757438 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21757408 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10488747 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10488738 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10488742 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10488746 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10488743 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10488744 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10488739 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10488740 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10488741 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10488735 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10488737 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10488736 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10488734 | - | - |
| Mouse Genome 430 2.0 Array | 1429912_at | - | - |
| Mouse Genome 430 2.0 Array | 1429912_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 76407_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 76407_PM_at | - | - |
| Mouse Expression Set 430 | 1429912_PM_at | - | - |
| Mouse Expression Set 430 | 1429912_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 419144 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 4540168 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004540168 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2723751 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2723751 | - | - |
| mouse-6 v1.1 (Illumina) | scl18477.12.1_53-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4540168 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl18477.12.1_53-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4540168 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3106706 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3033086 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2723751 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2723751 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3106706 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3033086 | - | - |

Sun5
Genomic Map