Assay Details
| Gene Symbol: Sun5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK006225.1 | - | 2 - 3 | 192 | 84 | |
| AY307077.1 | - | 2 - 3 | 211 | 84 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: Sun5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK006225.1 | - | 2 - 3 | 192 | 84 | |
| AY307077.1 | - | 2 - 3 | 211 | 84 | ||
Set Membership: |
|