Assay Details
| Gene Symbol: Sun5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK006225.1 | - | 8 - 9 | 595 | 126 | |
| AY307077.1 | - | 8 - 9 | 614 | 126 | ||
| FJ667498.1 | - | 9 - 10 | 603 | 126 | ||
More Information
Set Membership: |
|

Sun5
Genomic Map