Assay Details
| Gene Symbol: Vps37a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033560.3 | NP_291038.2 | 1 - 2 | 447 | 94 | |
| GenBank mRNA | AK016385.1 | - | 1 - 2 | 429 | 94 | |
| AK048942.1 | - | 1 - 2 | 427 | 94 | ||
| AK054055.1 | - | 1 - 2 | 429 | 94 | ||
| AK137479.1 | - | 1 - 2 | 432 | 94 | ||
| AK157507.1 | - | 1 - 2 | 413 | 94 | ||
| AK170280.1 | - | 1 - 2 | 416 | 94 | ||
| AK170943.1 | - | 1 - 2 | 405 | 94 | ||
| AK170961.1 | - | 1 - 2 | 436 | 94 | ||
| AK171055.1 | - | 1 - 2 | 436 | 94 | ||
| AK171221.1 | - | 1 - 2 | 448 | 94 | ||
| BC039290.1 | - | 1 - 2 | 403 | 5066508 | 94 | |
| BC110396.1 | - | 1 - 2 | 362 | 5253683 | 94 | |
More Information
Updated Mapping Information
Set Membership: |
|

Vps37a
Genomic Map