Assay Details
| Gene Symbol: Kcnh7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133207.2 | NP_573470.2 | 4 - 5 | 1100 | 101 | |
| GenBank mRNA | AJ291608.1 | - | 4 - 5 | 907 | 101 | |
| AK034003.1 | - | 4 - 5 | 1101 | 101 | ||
More Information
Set Membership: |
|

Kcnh7
Genomic Map